Methyl 5-bromo-2-chlorobenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 5-bromo-2-chlorobenzoate
OR52912-100G OR52912-25G
APOSOR52912-100GEA 513 EUR
APOSOR52912-100G APOSOR52912-25G
Methyl 5-bromo-2-chlorobenzoate 98%
Methyl 5-bromo-2-chlorobenzoate
Formula: C₈H₆BrClO₂
MW: 249.4890 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144770
CAS Number: 251085-87-7