Ethyl-(S)-(-)-lactate 99%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
OR5301-500G
APOSOR5301-500GEA 43.5 EUR
APOSOR5301-500G
Ethyl-(S)-(-)-lactate 99%
Ethyl (S)-(-)-lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 152…155 °C
Melting Pt: –25 °C
Density: 1.036 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
UN: 1993
ADR: 3,III