5-Bromo-8-quinolinol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromoquinolin-8-ol
OR53034-10G OR53034-25G OR53034-5G
APOSOR53034-10GEA 201 EUR
APOSOR53034-10G APOSOR53034-25G APOSOR53034-5G
5-Bromo-8-quinolinol 95%
5-Bromo-8-quinolinol
Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00457011
CAS Number: 1198-14-7
EINECS: 214-829-2