Methyl-3-hydroxybenzoate 99%
Supplier: Apollo Scientific
|
Warning
|
OR5305-1KG
OR5305-250G
OR5305-500G
APOSOR5305-1KGEA
290
EUR
APOSOR5305-1KG
APOSOR5305-250G
APOSOR5305-500G
Methyl-3-hydroxybenzoate 99%
Methyl 3-hydroxybenzoate
Formula:
C₈H₈O₃ Boiling Pt: 280…281 °C/708mmHg Melting Pt: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002295 CAS Number: 19438-10-9 EINECS: 243-071-5 |