4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid
Supplier: Apollo Scientific
Warning
OR53050-1G OR53050-250MG
APOSOR53050-1GEA 198 EUR
APOSOR53050-1G APOSOR53050-250MG
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid
Formula: C₇H₃ClN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11846188
CAS Number: 875515-76-7