3-Hydroxybenzhydrazide
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxybenzoic hydrazide, 3-Hydroxybenzoic acid hydrazide, 3-Hydroxybenzohydrazide, 3-Hydroxybenzene-1-carbohydrazide
OR53082-1G OR53082-25G OR53082-5G
APOSOR53082-1GEA 21.9 EUR
APOSOR53082-1G APOSOR53082-25G APOSOR53082-5G
3-Hydroxybenzhydrazide
3-Hydroxybenzhydrazide
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00014759
CAS Number: 5818-06-4
EINECS: 227-389-1