N-Ethylbenzylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: N-Benzylethylamine, N-Benzylethanamine
OR5309-100ML OR5309-25ML OR5309-500ML
APOSOR5309-100MLEA 81.5 EUR
APOSOR5309-100ML APOSOR5309-25ML APOSOR5309-500ML
N-Ethylbenzylamine 97%
N-Ethylbenzylamine
Formula: C₆H₅CH₂NHCH₂CH₃
MW: 135.21 g/mol
Boiling Pt: 191…194 °C/760mmHg
Density: 0.909 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00009031
CAS Number: 14321-27-8
UN: 2735
ADR: 8,II