EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
Supplier: Apollo Scientific
Danger
Synonyms: EDAC, WSC, EDC, N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine
OR53094-100G OR53094-25G OR53094-5G
APOSOR53094-100GEA 203 EUR
APOSOR53094-100G APOSOR53094-25G APOSOR53094-5G
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide
Formula: C₈H₁₇N₃
MW: 155.24 g/mol
Density: 0.877 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00044916
CAS Number: 1892-57-5
UN: 2735
ADR: 8,III