1,6-Dibromopyrene 98%
Supplier: Apollo Scientific
Warning
OR53108-25G OR53108-5G
APOSOR53108-25GEA 132 EUR
APOSOR53108-25G APOSOR53108-5G
1,6-Dibromopyrene 98%
1,6-Dibromopyrene
Formula: C₁₆H₈Br₂
MW: 360.05 g/mol
Melting Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263746
CAS Number: 27973-29-1