5-Chloro-2-methoxybenzaldehyde
Supplier: Apollo Scientific
Warning
OR53121-25G OR53121-5G
APOSOR53121-25GEA 134 EUR
APOSOR53121-25G APOSOR53121-5G
5-Chloro-2-methoxybenzaldehyde
5-Chloro-2-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.60 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00033854
CAS Number: 7035-09-8