Sebacic acid
Supplier: Apollo Scientific
Warning
Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid
OR53173-250G OR53173-100G OR53173-500G
APOSOR53173-250GEA 55.3 EUR
APOSOR53173-250G APOSOR53173-100G APOSOR53173-500G
Sebacic acid
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 295 °C/100mmHg
Melting Pt: 131…134 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6