4-tert-Butyl-2-methylthiazole
Supplier: Apollo Scientific
OR5322-25G OR5322-5G OR5322-1G OR5322-10G
APOSOR5322-25GEA 0 EUR
APOSOR5322-25G APOSOR5322-5G APOSOR5322-1G APOSOR5322-10G
4-tert-Butyl-2-methylthiazole
4-tert-Butyl-2-methylthiazole
Formula: C₈H₁₃NS
Boiling Pt: 174…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051643
CAS Number: 15679-11-5