3-Bromobenzoic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: m-Bromobenzoic Acid
OR5328-100G OR5328-25G
APOSOR5328-100GEA 114 EUR
APOSOR5328-100G APOSOR5328-25G
3-Bromobenzoic acid 98%
3-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002487
CAS Number: 585-76-2
EINECS: 209-562-3