2,4,6-Trichlorobenzoic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,6-TBA
OR5333-100G OR5333-250G OR5333-25G OR5333-500G
APOSOR5333-100GEA 89.9 EUR
APOSOR5333-100G APOSOR5333-250G APOSOR5333-25G APOSOR5333-500G
2,4,6-Trichlorobenzoic acid 99%
2,4,6-Trichlorobenzoic acid
Formula: C₇H₃Cl₃O₂
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00060699
CAS Number: 50-43-1