2-Chlorobenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: o-Chlorobenzoyl chloride
OR5336-100G OR5336-25G
APOSOR5336-100GEA 36.4 EUR
APOSOR5336-100G APOSOR5336-25G
2-Chlorobenzoyl chloride 97%
2-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 237…239 °C
Melting Pt: –1 °C
Density: 1.38 g/cm³ (25 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00000660
CAS Number: 609-65-4
EINECS: 210-194-0
UN: 3265
ADR: 8,II