3,4-Dibromobenzaldehyde 99%
Supplier: Apollo Scientific
Warning
OR5345-1G OR5345-5G OR5345-25G
APOSOR5345-1GEA 21.9 EUR
APOSOR5345-1G APOSOR5345-5G APOSOR5345-25G
3,4-Dibromobenzaldehyde 99%
3,4-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
MW: 263.92 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00236033
CAS Number: 74003-55-7