3,5-Lutidine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 3,5-Dimethylpyridine
OR5347-1KG OR5347-250G OR5347-5KG
APOSOR5347-1KGEA 132 EUR
APOSOR5347-1KG APOSOR5347-250G APOSOR5347-5KG
3,5-Lutidine 99%
3,5-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 169…170 °C
Melting Pt: –7 °C
Density: 0.941 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00006404
CAS Number: 591-22-0
EINECS: 209-708-6
UN: 1992
ADR: 3+6.1,III