3-Methylbutanone 98%
Supplier: Apollo Scientific
Danger
Synonyms: Isopropyl Methyl Ketone, 3-Methyl-2-butanone
OR5350-250ML
APOSOR5350-250MLEA 87 EUR
APOSOR5350-250ML
3-Methylbutanone 98%
3-Methylbutanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 94…95 °C
Melting Pt: –92 °C
Density: 0.805 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008919
CAS Number: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II