m-Toluidine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline
OR5362-250G OR5362-1KG
APOSOR5362-250GEA 87 EUR
APOSOR5362-250G APOSOR5362-1KG
m-Toluidine 99%
m-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 203…204 °C
Melting Pt: –30 °C
Density: 0.999 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007808
CAS Number: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1 II
Merck Index: 13,09614