(4-Chlorophenyl)acetonitrile 98+%
Supplier: Apollo Scientific
Danger
OR5364-1KG OR5364-500G OR5364-5KG
APOSOR5364-1KGEA 36.4 EUR
APOSOR5364-1KG APOSOR5364-500G APOSOR5364-5KG
(4-Chlorophenyl)acetonitrile 98+%
(4-Chlorophenyl)acetonitrile
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 25…31 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00001918
CAS Number: 140-53-4
EINECS: 205-418-9
UN: 3439
ADR: 6.1,II