2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95+%
Supplier: Apollo Scientific
Danger
Synonyms: Bis(diisopropylamino)(2-cyanoethoxy)phosphine, 3-((Bis(diisopropylamino)phosphino)oxy)propanenitrile
OR5367-5G OR5367-10G OR5367-1G
APOSOR5367-5GEA 24.8 EUR
APOSOR5367-5G APOSOR5367-10G APOSOR5367-1G
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95+%
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite
Formula: C₁₅H₃₂N₃OP
MW: 301.41 g/mol
Boiling Pt: 100 °C/0.5mmHg
Density: 0.949 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Freezer
MDL Number: MFCD00012213
CAS Number: 102691-36-1
UN: 1993
ADR: 3 III