2-Amino-1,3-propanediol 98%
Supplier: Apollo Scientific
Danger
Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol
OR5368-100G OR5368-25G OR5368-500G
APOSOR5368-100GEA 53.9 EUR
APOSOR5368-100G APOSOR5368-25G APOSOR5368-500G
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 128 °C/0.8mmHg
Melting Pt: 52…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
UN: 3259
ADR: 8 III