2-Nitrobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Nitrobenzene-1-sulphonyl chloride
OR5376-100G OR5376-25G OR5376-500G
APOSOR5376-100GEA 69.6 EUR
APOSOR5376-100G APOSOR5376-25G APOSOR5376-500G
2-Nitrobenzenesulphonyl chloride 97%
2-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
MW: 221.62 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00007430
CAS Number: 1694-92-4
EINECS: 216-907-1
UN: 3261
ADR: 8,II