2,5-Dichlorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Dichlorobenzenesulphonyl Chloride, 2,5-Dichlorobenzene-1-sulphonyl chloride
OR5382-100G OR5382-25G
APOSOR5382-100GEA 197 EUR
APOSOR5382-100G APOSOR5382-25G
2,5-Dichlorobenzenesulphonyl chloride 98%
2,5-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Melting Pt: 36…37 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007429
CAS Number: 5402-73-3
EINECS: 226-453-6
UN: 3261
ADR: 8,II