2,6-Dichlorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR5383-5G OR5383-100G OR5383-25G
APOSOR5383-5GEA 21.9 EUR
APOSOR5383-5G APOSOR5383-100G APOSOR5383-25G
2,6-Dichlorobenzenesulphonyl chloride
2,6-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052311
CAS Number: 6579-54-0
UN: 3261
ADR: 8,III