3-Cyano-4-methylaniline
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-2-methylbenzonitrile
OR5392-25G OR5392-5G OR5392-1G
APOSOR5392-25GEA 147 EUR
APOSOR5392-25G APOSOR5392-5G APOSOR5392-1G
3-Cyano-4-methylaniline
3-Cyano-4-methylaniline
Formula: C₈H₈N₂
Boiling Pt: 110 °C/22mmHg
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00017608
CAS Number: 50670-64-9