1,4-Naphthoquinone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
OR5393-2.5KG OR5393-5KG
APOSOR5393-2.5KGEA 65.4 EUR
APOSOR5393-2.5KG APOSOR5393-5KG
1,4-Naphthoquinone 97%
1,4-Naphthoquinone
Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2811
ADR: 6.1,I