Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(R)-(+)-3-Boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, (R)-(+)-3-(tert-Butoxycarbonyl)-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, Methyl (R)-N-Boc-2,2-dimethyloxazolidine-4-carboxylate, (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylic acid 3-tert-Butyl 4-Methyl ester, 3-tert-Butyl 4-Methyl (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylate
OR5403-10G
OR5403-1G
OR5403-5G
APOSOR5403-10GEA
248
EUR
APOSOR5403-10G
APOSOR5403-1G
APOSOR5403-5G
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate
Formula:
C₁₂H₂₁NO₅ Boiling Pt: 89 °C/0.8mmHg Density: 1.082 g/cm³ (25 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674041 CAS Number: 95715-86-9 |