1-Boc-4-hydroxypiperidine 98+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate
OR5404-100G
OR5404-25G
OR5404-500G
APOSOR5404-100GEA
85.9
EUR
APOSOR5404-100G
APOSOR5404-25G
APOSOR5404-500G
1-Boc-4-hydroxypiperidine 98+%
1-Boc-4-hydroxypiperidine
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075174 CAS Number: 109384-19-2 |