1-Chloro-4-nitrobenzene 98+%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Chloronitrobenzene
OR5413-1KG OR5413-250G OR5413-5KG
APOSOR5413-1KGEA 84.1 EUR
APOSOR5413-1KG APOSOR5413-250G APOSOR5413-5KG
1-Chloro-4-nitrobenzene 98+%
1-Chloro-4-nitrobenzene
Formula: C₆H₄ClNO₂
Boiling Pt: 242 °C
Melting Pt: 80…83 °C
Density: 1.298 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007285
CAS Number: 100-00-5
UN: 1578
ADR: 6.1,II