3-Iodophenol 97%
Supplier: Apollo Scientific
Warning
OR5426-25G OR5426-100G OR5426-5G OR5426-1G
APOSOR5426-25GEA 109 EUR
APOSOR5426-25G APOSOR5426-100G APOSOR5426-5G APOSOR5426-1G
3-Iodophenol 97%
3-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 190 °C/97.5mmHg
Melting Pt: 42…44 °C
Flash Pt: >113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2