4-Chlorothiophenol 99%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol
OR54321-100G OR54321-500G OR54321-25G
APOSOR54321-100GEA 50.9 EUR
APOSOR54321-100G APOSOR54321-500G APOSOR54321-25G
4-Chlorothiophenol 99%
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205…207 °C
Melting Pt: 48…52 °C
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 3261
ADR: 8,II