2,4-Dimethylimidazole 98%
Supplier: Apollo Scientific
Danger
OR54335-5G OR54335-100G OR54335-25G
APOSOR54335-5GEA 21.9 EUR
APOSOR54335-5G APOSOR54335-100G APOSOR54335-25G
2,4-Dimethylimidazole 98%
2,4-Dimethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 266 °C/733mmHg
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00022365
CAS Number: 930-62-1
EINECS: 213-221-4
UN: 3263
ADR: 8 III