3-Chlorophthalic anhydride, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloroisobenzofuran-1,3-dione
OR54359-25G OR54359-5G OR54359-100G
APOSOR54359-25GEA 21.9 EUR
APOSOR54359-25G APOSOR54359-5G APOSOR54359-100G
3-Chlorophthalic anhydride, Technical Grade
3-Chlorophthalic anhydride
Formula: C₈H₃ClO₃
MW: 182.56 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00023107
CAS Number: 117-21-5
EINECS: 204-179-8