2',6'-Dichloroacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,6-Dichlorophenyl)ethanone
OR5436-1G OR5436-25G OR5436-5G OR5436-10G
APOSOR5436-1GEA 21.9 EUR
APOSOR5436-1G APOSOR5436-25G APOSOR5436-5G APOSOR5436-10G
2',6'-Dichloroacetophenone
2',6'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 176…180 °C/83mmHg
Melting Pt: 37…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00016339
CAS Number: 2040-05-3
EINECS: 218-035-7