1-Methyl-1H-pyrazole-4-sulphonyl chloride
Supplier: Apollo Scientific
Danger
OR54372-1G OR54372-5G OR54372-250MG
APOSOR54372-1GEA 26.1 EUR
APOSOR54372-1G APOSOR54372-5G APOSOR54372-250MG
1-Methyl-1H-pyrazole-4-sulphonyl chloride
1-Methyl-1H-pyrazole-4-sulphonyl chloride
Formula: C₄H₅ClN₂O₂S
MW: 179.98 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Freezer
MDL Number: MFCD04968667
CAS Number: 288148-34-5
UN: 3261
ADR: 8 III