3,6-Diiodocarbazole
Supplier: Apollo Scientific
Warning
Synonyms: 3,6-Diiodo-9H-carbazole
OR54455-100G OR54455-25G OR54455-5G
APOSOR54455-100GEA 120 EUR
APOSOR54455-100G APOSOR54455-25G APOSOR54455-5G
3,6-Diiodocarbazole
3,6-Diiodocarbazole
Formula: C₁₂H₇I₂N
MW: 419.00 g/mol
Melting Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00450017
CAS Number: 57103-02-3