2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(4,6-Dimethylpyrimidin-2-yl)thio]aniline
OR5446-1G OR5446-500MG
APOSOR5446-1GEA 105 EUR
APOSOR5446-1G APOSOR5446-500MG
2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine
2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine
Formula: C₁₂H₁₃N₃S
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00203030
CAS Number: 102243-12-9