(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Supplier: Apollo Scientific
Warning
Synonyms: (S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl
OR54464-100G OR54464-25G OR54464-5G
APOSOR54464-100GEA 398 EUR
APOSOR54464-100G APOSOR54464-25G APOSOR54464-5G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223