Methyl acetoacetate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-oxobutanoate
OR54474-1KG OR54474-5KG
APOSOR54474-1KGEA 109 EUR
APOSOR54474-1KG APOSOR54474-5KG
Methyl acetoacetate
Methyl acetoacetate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 170 °C
Melting Pt: –180 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008784
CAS Number: 105-45-3
Merck Index: 12,06090