7-Methyl-1-tetralone
Supplier: Apollo Scientific
Warning
OR54530-1G OR54530-5G
APOSOR54530-1GEA 24.4 EUR
APOSOR54530-1G APOSOR54530-5G
7-Methyl-1-tetralone
7-Methyl-1-tetralone
Formula: C₁₁H₁₂O
Boiling Pt: 115…120 °C/5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00087600
CAS Number: 22009-37-6