Methyl 1-Oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 1-Oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid methyl ester, Methyl 1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate
OR54535-1G OR54535-5G
APOSOR54535-1GEA 43.5 EUR
APOSOR54535-1G APOSOR54535-5G
Methyl 1-Oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate
Methyl 1-Oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate
Formula: C₁₂H₁₂O₃
MW: 204.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00667560
CAS Number: 7442-52-6