2,4-Dichloropyrimidine 97%
Supplier: Apollo Scientific
Warning
OR5465-250G OR5465-100G OR5465-25G
APOSOR5465-250GEA 102 EUR
APOSOR5465-250G APOSOR5465-100G APOSOR5465-25G
2,4-Dichloropyrimidine 97%
2,4-Dichloropyrimidine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 101 °C/23mmHg
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6