1,2-Bis(2-chloroethoxy)ethane 97%
Supplier: Apollo Scientific
Danger
OR54670-100G OR54670-500G OR54670-25G
APOSOR54670-100GEA 46.4 EUR
APOSOR54670-100G APOSOR54670-500G APOSOR54670-25G
1,2-Bis(2-chloroethoxy)ethane 97%
1,2-Bis(2-chloroethoxy)ethane
Formula: ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MW: 187.07 g/mol
Boiling Pt: 235 °C
Density: 1.197 g/cm³ (25 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00000976
CAS Number: 112-26-5
UN: 2810
ADR: 6.1,III