2-Methyl-5-isopropylaniline
Supplier: Apollo Scientific
Warning
Synonyms: 5-Isopropyl-2-methylaniline, 2-Amino-p-cymene, 5-Isopropyl-o-toluidine
OR54679-1G OR54679-5G OR54679-25G
APOSOR54679-1GEA 21.9 EUR
APOSOR54679-1G APOSOR54679-5G APOSOR54679-25G
2-Methyl-5-isopropylaniline
2-Methyl-5-isopropylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Density: 0.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044503
CAS Number: 2051-53-8