3,5-Dichlorobenzaldehyde 98%
Supplier: Apollo Scientific
OR54707-100G OR54707-25G
APOSOR54707-100GEA 288 EUR
APOSOR54707-100G APOSOR54707-25G
3,5-Dichlorobenzaldehyde 98%
3,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 235…240 °C
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00003352
CAS Number: 10203-08-4
UN: 3261
ADR: 8,II