3-(Dimethylamino)-1-propanol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Dimethylamino)propan-1-ol, 3-Dimethylamino-1-propanol
OR5472-100G OR5472-25G OR5472-5G
APOSOR5472-100GEA 319 EUR
APOSOR5472-100G APOSOR5472-25G APOSOR5472-5G
3-(Dimethylamino)-1-propanol 98%
3-(Dimethylamino)-1-propanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 163…164 °C
Melting Pt: –35 °C
Density: 0.88 g/cm³ (25 °C)
Flash Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00002945
CAS Number: 3179-63-3
UN: 2734
ADR: 8+3 II