Maleimide
Supplier: Apollo Scientific
Danger
OR5487-100G OR5487-25G
APOSOR5487-100GEA 101 EUR
APOSOR5487-100G APOSOR5487-25G
Maleimide
Maleimide
Formula: C₄H₃NO₂
Boiling Pt: 97…103 °C/5mmHg
Melting Pt: 91…93 °C
Density: 1.249 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8+6.1,II