4-Bromo-1,2-dichlorobenzene 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3,4-dichlorobenzene
OR5488-100G OR5488-250G OR5488-25G OR5488-500G
APOSOR5488-100GEA 24.8 EUR
APOSOR5488-100G APOSOR5488-250G APOSOR5488-25G APOSOR5488-500G
4-Bromo-1,2-dichlorobenzene 98+%
4-Bromo-1,2-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140…142 °C/30mmHg
Melting Pt: 22…25 °C
Density: 1.781 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6