5-Azaindole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine
OR5493-1G OR5493-5G OR5493-25G
APOSOR5493-1GEA 21.9 EUR
APOSOR5493-1G APOSOR5493-5G APOSOR5493-25G
5-Azaindole 98%
5-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 132 °C/0.22mmHg
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00955936
CAS Number: 271-34-1